Gene Transcripts
- Gene Symbol
- Klra8
- Entrez Gene ID
- 16639
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 8
- Gene Aliases
- Cmv-1, Cmv1, Ly49h, Ly49u<129>
- Chromosome Location
- Chr. 6: 130115226 - 130129898 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
- Gene Symbol
- Klra14-ps
- Entrez Gene ID
- 654449
- Gene Name
- killer cell lectin-like receptor subfamily A, member 14, pseudogene
- Gene Aliases
- EG654449, Gm15858, Klra14, Klra29, Klra31, Ly49N, OTTMUSG00000026959
- Chromosome Location
- Chr. 6: 130149106 - 130160748 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY860969.1 |
- |
|
695 |
|
- Gene Symbol
- Klra21
- Entrez Gene ID
- 93968
- Gene Name
- killer cell lectin-like receptor subfamily A, member 21
- Gene Aliases
- Klra-ps1, Ly49H, Ly49u
- Chromosome Location
- Chr. 6: 130082598 - 130096486 on Build GRCm38
- UniGene ID
- Mm.439698
- Species
- Mus musculus
- Gene Symbol
- Klra3
- Entrez Gene ID
- 16634
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 3
- Gene Aliases
- 5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
- Chromosome Location
- Chr. 6: 130323289 - 130337626 on Build GRCm38
- UniGene ID
- Mm.425338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC141217.1 |
- |
|
788 |
|
- Gene Symbol
- Klra9
- Entrez Gene ID
- 16640
- Gene Name
- killer cell lectin-like receptor subfamily A, member 9
- Gene Aliases
- LY49I1, Ly49I
- Chromosome Location
- Chr. 6: 130178626 - 130194374 on Build GRCm38
- UniGene ID
- Mm.457996
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U49867.1 |
- |
|
858 |
|
View Full Product Details