Gene Transcripts

Gene Symbol
Rab23
Entrez Gene ID
19335
Gene Name
RAB23, member RAS oncogene family
Gene Aliases
AW545388, opb, opb2
Chromosome Location
Chr. 1: 33719896 - 33742564 on Build GRCm38
UniGene ID
Mm.86744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159729.1 NP_001153201.1 5 709
NM_008999.4 NP_033025.2 5 821
GenBank mRNA AB232618.1 - 3 386
AK016493.1 - 5 721
AK019666.1 - 2 270
AK045454.1 - 5 580
AK047068.1 - 5 822
AK082017.1 - 5 785
AK164057.1 - 5 905
BC025578.1 - 5 716 5323566
Z22821.1 - 3 406

View Full Product Details