Gene Transcripts

Gene Symbol
Ttc23
Entrez Gene ID
67009
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
1600012K10Rik
Chromosome Location
Chr. 7: 67647410 - 67730060 on Build GRCm38
UniGene ID
Mm.249356
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168475.1 NP_001161947.1 6 1077
NM_001168476.1 NP_001161948.1 5 877
NM_025905.3 NP_080181.2 6 1077
GenBank mRNA AK005431.1 - 5 838
AK076802.1 - 5 671
AK086049.1 - 1144
AK141754.1 - 6 1063
AK207178.1 - 3 275
BC038179.1 - 6 1048 4485073

View Full Product Details