Gene Transcripts

Gene Symbol
Sgms2
Entrez Gene ID
74442
Gene Name
sphingomyelin synthase 2
Gene Aliases
4933405A16Rik, 5133401H06Rik, AI854299
Chromosome Location
Chr. 3: 131318985 - 131491192 on Build GRCm38
UniGene ID
Mm.273360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028943.5 NP_083219.2 2 660
GenBank mRNA AK016659.1 - 2 674
BC117782.2 - 2 664 40092081

View Full Product Details