Gene Transcripts

Gene Symbol
Rsph9
Entrez Gene ID
75564
Gene Name
radial spoke head 9 homolog (Chlamydomonas)
Gene Aliases
1700027N10Rik
Chromosome Location
Chr. 17: 46122038 - 46144237 on Build GRCm38
UniGene ID
Mm.17993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029338.3 NP_083614.1 3 559
GenBank mRNA AK006440.1 - 3 562
AK140720.1 - 1 40
AK165820.1 - 3 430
BC019423.1 - 3 533 4922914

View Full Product Details