Gene Transcripts

Gene Symbol
Tiparp
Entrez Gene ID
99929
Gene Name
TCDD-inducible poly(ADP-ribose) polymerase
Gene Aliases
ARTD14, AW558171, PARP7
Chromosome Location
Chr. 3: 65528447 - 65555518 on Build GRCm38
UniGene ID
Mm.246398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178892.5 NP_849223.2 2 743
GenBank mRNA AK028529.1 - 2 744
AK032453.1 - 2 727
AK150248.1 - 2 725
AK162094.1 - 2 749
AK163347.1 - 2 783
BC068173.1 - 2 717 30616032

View Full Product Details