Gene Transcripts

Gene Symbol
Hs3st3a1
Entrez Gene ID
15478
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1
Gene Aliases
3Ost3a, Hs3st3a
Chromosome Location
Chr. 11: 64435332 - 64522835 on Build GRCm38
UniGene ID
Mm.340214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178870.5 NP_849201.1 2 2209
GenBank mRNA AK032282.1 - 2 1177
AK051284.1 - 2 2210
BC094320.1 - 2 2263 6400013

View Full Product Details