Gene Transcripts

Gene Symbol
Slc25a20
Entrez Gene ID
57279
Gene Name
solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20
Gene Aliases
1110007P09Rik, C78826, Cact, mCAC
Chromosome Location
Chr. 9: 108662098 - 108684641 on Build GRCm38
UniGene ID
Mm.29666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020520.4 NP_065266.1 3, 4 453
GenBank mRNA AK075624.1 - 3, 4 454
AK152436.1 - 3, 4 424
BC029733.1 - 3, 4 450 4019406
BC052871.1 - 3, 4 376 30032324

View Full Product Details