Gene Transcripts

Gene Symbol
Selenbp2
Entrez Gene ID
20342
Gene Name
selenium binding protein 2
Gene Aliases
AP56, Lpsb2
Chromosome Location
Chr. 3: 94693573 - 94704406 on Build GRCm38
UniGene ID
Mm.225405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019414.2 NP_062287.1 12 1676
GenBank mRNA BC024106.1 - 12 1674
S56599.1 - 12 1676

View Full Product Details