Gene Transcripts
- Gene Symbol
- Vmn1r81
- Entrez Gene ID
- 171244
- Gene Name
- vomeronasal 1 receptor 81
- Gene Aliases
- V1rg9
- Chromosome Location
- Chr. 7: 12259759 - 12260679 on Build GRCm38
- UniGene ID
- Mm.377200
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134210.1 |
NP_598971.1
|
1 |
209 |
|
View Full Product Details