Gene Transcripts
- Gene Symbol
- NRBP1
- Entrez Gene ID
- 29959
- Gene Name
- nuclear receptor binding protein 1
- Gene Aliases
- BCON3, MADM, MUDPNP, NRBP
- Chromosome Location
- Chr. 2: 27427790 - 27442259 on Build GRCh38
- UniGene ID
- Hs.515876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321357.1 |
NP_001308286.1
|
5, 6 |
1269 |
|
NM_001321358.1 |
NP_001308287.1
|
4, 5 |
642 |
|
NM_001321359.1 |
NP_001308288.1
|
4, 5 |
637 |
|
NM_001321361.1 |
NP_001308290.1
|
4, 5 |
642 |
|
NM_001321362.1 |
NP_001308291.1
|
4, 5 |
642 |
|
NM_013392.3 |
NP_037524.1
|
4, 5 |
637 |
|
GenBank mRNA |
AF113249.1 |
- |
4, 5 |
549 |
|
AK001946.1 |
- |
4, 5 |
527 |
|
AK027538.1 |
- |
4, 5 |
545 |
|
AK122664.1 |
- |
5, 6 |
1269 |
|
AK223136.1 |
- |
4, 5 |
533 |
|
AK225540.1 |
- |
4, 5 |
563 |
|
AK301345.1 |
- |
5, 6 |
480 |
|
AL136682.1 |
- |
4, 5 |
524 |
|
BC001221.1 |
- |
4, 5 |
551 |
3534543 |
BQ949042.1 |
- |
4, 5 |
583 |
|
View Full Product Details