Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002838.3 | NP_001002838.1 | 20 | 4515 | |
NM_020922.4 | NP_065973.2 | 20 | 4656 | ||
XM_006724593.3 | XP_006724656.1 | 20 | 4453 | ||
XM_011530802.2 | XP_011529104.1 | 20 | 4453 | ||
XM_017029741.1 | XP_016885230.1 | 20 | 4595 | ||
XM_017029742.1 | XP_016885231.1 | 20 | 4672 | ||
XM_017029743.1 | XP_016885232.1 | 20 | 4587 | ||
XM_017029744.1 | XP_016885233.1 | 20 | 4489 | ||
XM_017029745.1 | XP_016885234.1 | 20 | 4594 | ||
GenBank mRNA | AB046786.1 | - | 14 | 2787 | |
AJ409088.2 | - | 19 | 4076 | ||
AY082340.1 | - | 19 | 4217 | ||
AY352048.1 | - | 20 | 4656 |