Gene Transcripts
- Gene Symbol
- MAPK11
- Entrez Gene ID
- 5600
- Gene Name
- mitogen-activated protein kinase 11
- Gene Aliases
- P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta
- Chromosome Location
- Chr. 22: 50263713 - 50270393 on Build GRCh38
- UniGene ID
- Hs.57732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002751.6 |
NP_002742.3
|
8, 9 |
786 |
|
NR_110887.1 |
|
8, 9 |
786 |
|
XM_011530706.1 |
XP_011529008.1
|
6, 7 |
510 |
|
GenBank mRNA |
AF001008.1 |
- |
8, 9 |
685 |
|
AF001174.1 |
- |
8, 9 |
691 |
|
AF031135.1 |
- |
8, 9 |
700 |
|
AK291845.1 |
- |
8, 9 |
737 |
|
AK308969.1 |
- |
8, 9 |
702 |
|
BC027933.1 |
- |
8, 9 |
743 |
5199628 |
BX361670.2 |
- |
8, 9 |
769 |
|
BX444192.2 |
- |
8, 9 |
784 |
|
CR456514.1 |
- |
8, 9 |
687 |
|
U53442.1 |
- |
8, 9 |
728 |
|
U92268.1 |
- |
8, 9 |
689 |
|
Y14440.1 |
- |
8, 9 |
691 |
|
View Full Product Details