Gene Transcripts
- Gene Symbol
- PTPDC1
- Entrez Gene ID
- 138639
- Gene Name
- protein tyrosine phosphatase domain containing 1
- Gene Aliases
- PTP9Q22
- Chromosome Location
- Chr. 9: 94030591 - 94109856 on Build GRCh38
- UniGene ID
- Hs.437943
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253829.1 |
NP_001240758.1
|
5, 6 |
864 |
|
NM_001253830.1 |
NP_001240759.1
|
6, 7 |
925 |
|
NM_152422.4 |
NP_689635.3
|
5, 6 |
858 |
|
NM_177995.2 |
NP_818931.1
|
6, 7 |
940 |
|
XM_005251711.4 |
XP_005251768.1
|
6, 7 |
2882 |
|
XM_011518240.2 |
XP_011516542.1
|
6, 7 |
690 |
|
XM_011518241.2 |
XP_011516543.1
|
6, 7 |
644 |
|
XM_017014280.1 |
XP_016869769.1
|
7, 8 |
749 |
|
XM_017014281.1 |
XP_016869770.1
|
6, 7 |
668 |
|
XM_017014282.1 |
XP_016869771.1
|
3, 4 |
771 |
|
GenBank mRNA |
AK124912.1 |
- |
5, 6 |
744 |
|
AK289692.1 |
- |
5, 6 |
815 |
|
AK310403.1 |
- |
5, 6 |
822 |
|
AY171233.1 |
- |
6, 7 |
945 |
|
BC067120.1 |
- |
6, 7 |
925 |
5539182 |
DA312531.1 |
- |
4, 5 |
440 |
|
View Full Product Details