Gene Transcripts
- Gene Symbol
- UBAC2
- Entrez Gene ID
- 337867
- Gene Name
- UBA domain containing 2
- Gene Aliases
- PHGDHL1
- Chromosome Location
- Chr. 13: 99200425 - 99386499 on Build GRCh38
- UniGene ID
- Hs.508545
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001144072.1 |
NP_001137544.1
|
7, 8 |
1299 |
|
NM_177967.3 |
NP_808882.1
|
5, 6 |
1213 |
|
NR_026644.1 |
|
7, 8 |
1443 |
|
XM_006719948.3 |
XP_006720011.1
|
8, 9 |
1009 |
|
XM_011521082.2 |
XP_011519384.1
|
5, 6 |
1069 |
|
XM_011521083.2 |
XP_011519385.1
|
6, 7 |
776 |
|
XM_011521084.2 |
XP_011519386.1
|
7, 8 |
864 |
|
XM_017020553.1 |
XP_016876042.1
|
2, 3 |
4091 |
|
GenBank mRNA |
AF339812.1 |
- |
2, 3 |
283 |
|
AK054563.1 |
- |
5, 6 |
1214 |
|
AK055110.1 |
- |
4, 5 |
996 |
|
AK075419.1 |
- |
7, 8 |
870 |
|
AK075516.1 |
- |
5, 6 |
640 |
|
AK124404.1 |
- |
7, 8 |
597 |
|
AK129861.1 |
- |
7, 8 |
898 |
|
AK290342.1 |
- |
7, 8 |
1299 |
|
AK300908.1 |
- |
7, 8 |
785 |
|
BC063559.1 |
- |
5, 6 |
658 |
|
BC072674.1 |
- |
3, 4 |
435 |
|
BC121138.1 |
- |
5, 6 |
870 |
|
BC121139.1 |
- |
7, 8 |
1042 |
40120987 |
View Full Product Details