Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114173.2 | NP_001107645.1 | 2 | 402 | |
NM_001814.5 | NP_001805.3 | 2 | 402 | ||
NM_148170.4 | NP_680475.1 | 2 | 402 | ||
GenBank mRNA | AF234263.1 | - | 2 | 313 | |
AF234264.1 | - | 2 | 313 | ||
AF254757.1 | - | 2 | 313 | ||
AF525032.1 | - | 2 | 313 | ||
AF525033.1 | - | 2 | 313 | ||
AK223038.1 | - | 2 | 351 | ||
AK292117.1 | - | 2 | 320 | ||
AK296190.1 | - | 3 | 300 | ||
AK311923.1 | - | 2 | 351 | ||
BC054028.1 | - | 2 | 341 | ||
BC100891.1 | - | 2 | 337 | ||
BC100892.1 | - | 2 | 337 | ||
BC100893.1 | - | 2 | 337 | ||
BC100894.1 | - | 2 | 337 | ||
BC109386.1 | - | 2 | 337 | ||
BC110071.1 | - | 2 | 337 | ||
BC113850.1 | - | 2 | 337 | ||
BC113897.1 | - | 2 | 337 | ||
BX380095.2 | - | 2 | 402 | ||
BX459226.2 | - | 2 | 386 | ||
BX537913.1 | - | 2 | 376 | ||
CB114425.1 | - | 2 | 383 | ||
X87212.1 | - | 2 | 320 |