Gene Transcripts
- Gene Symbol
- PON2
- Entrez Gene ID
- 5445
- Gene Name
- paraoxonase 2
- Gene Aliases
- Chromosome Location
- Chr. 7: 95404862 - 95435072 on Build GRCh38
- UniGene ID
- Hs.514420
- Species
- Homo sapiens
- Gene Symbol
- ZFHX4
- Entrez Gene ID
- 79776
- Gene Name
- zinc finger homeobox 4
- Gene Aliases
- ZFH4, ZHF4
- Chromosome Location
- Chr. 8: 76681219 - 76867285 on Build GRCh38
- UniGene ID
- Hs.458973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR933619.1 |
- |
|
5576 |
|
View Full Product Details