Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000499.4 | NP_000490.1 | 7 | 1952 | |
NM_001319216.1 | NP_001306145.1 | 6 | 1868 | ||
NM_001319217.1 | NP_001306146.1 | 7 | 1955 | ||
XM_017021953.1 | XP_016877442.1 | 8 | 1871 | ||
GenBank mRNA | AK310810.1 | - | 7 | 1999 | |
BC023019.1 | - | 7 | 1837 | 5123393 | |
K03191.1 | - | 7 | 1842 |