Gene Transcripts

Gene Symbol
ALOX15
Entrez Gene ID
246
Gene Name
arachidonate 15-lipoxygenase
Gene Aliases
12-LOX, 15-LOX-1, 15LOX-1
Chromosome Location
Chr. 17: 4630919 - 4641665 on Build GRCh38
UniGene ID
Hs.73809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001140.3 NP_001131.3 14 1852
GenBank mRNA AK290309.1 - 14 1865
AK296792.1 - 12 1509
AK296821.1 - 15 1937
AK296904.1 - 14 1748
AK312557.1 - 14 1865
AK316126.1 - 14 2008
BC029032.1 - 14 1852 5179700
M23892.1 - 14 1841

View Full Product Details