Gene Transcripts
- Gene Symbol
- NUP88
- Entrez Gene ID
- 4927
- Gene Name
- nucleoporin 88
- Gene Aliases
- Chromosome Location
- Chr. 17: 5384833 - 5419739 on Build GRCh38
- UniGene ID
- Hs.584784
- Species
- Homo sapiens
- Gene Symbol
- RABEP1
- Entrez Gene ID
- 9135
- Gene Name
- rabaptin, RAB GTPase binding effector protein 1
- Gene Aliases
- RAB5EP, RABPT5
- Chromosome Location
- Chr. 17: 5282263 - 5386340 on Build GRCh38
- UniGene ID
- Hs.584784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM678290.1 |
- |
1 |
202 |
|
View Full Product Details