Gene Transcripts

Gene Symbol
RBP3
Entrez Gene ID
5949
Gene Name
retinol binding protein 3
Gene Aliases
D10S64, D10S65, D10S66, IRBP, RBPI, RP66
Chromosome Location
Chr. 10: 47348371 - 47357875 on Build GRCh38
UniGene ID
Hs.591928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002900.2 NP_002891.1 4 3578
GenBank mRNA AB593121.1 - 4 3570
BC039844.1 - 4 3568 5578501
J03912.1 - 4 3461
M22453.1 - 4 3579

View Full Product Details