Gene Transcripts

Gene Symbol
MYB
Entrez Gene ID
4602
Gene Name
MYB proto-oncogene, transcription factor
Gene Aliases
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location
Chr. 6: 135180981 - 135219173 on Build GRCh38
UniGene ID
Hs.606320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130173.1 NP_001123645.1 8 1138
NM_001161658.1 NP_001155130.1 8 1138
NM_001161659.1 NP_001155131.1 8 1138
NM_005375.3 NP_005366.2 8 1138
NR_134960.1 8 1138
NR_134961.1 8 1138
NR_134963.1 8 1138
GenBank mRNA AF104863.1 - 8 1016
AJ606318.1 - 8 1138
AJ606319.1 - 8 1138
AJ606322.1 - 8 1138
AJ606323.1 - 8 1138
AJ606324.1 - 8 1138
AJ616235.1 - 8 1053
AY787443.1 - 8 939
AY787444.1 - 6 646
AY787450.1 - 8 939
AY787451.1 - 8 939
AY787452.1 - 8 939
AY787453.1 - 8 939
AY787464.1 - 8 939
AY787465.1 - 8 939
AY787466.1 - 8 939
AY787467.1 - 8 939
AY787468.1 - 8 939
AY787469.1 - 8 939
AY787470.1 - 8 939
AY787471.1 - 8 939
AY787472.1 - 8 939
AY787473.1 - 8 939
AY787474.1 - 8 939
AY787475.1 - 8 939
BC064955.1 - 8 1090 6069320
M13665.1 - 6 801
M15024.1 - 8 1052

View Full Product Details