Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr. 7: 128954180 - 129055173 on Build GRCh38
UniGene ID
Hs.193613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191028.2 NP_001177957.2 19, 20 2805
NM_012470.3 NP_036602.1 20, 21 2997
NR_034053.2 21, 22 3158
XM_011515989.2 XP_011514291.1 20, 21 2599
GenBank mRNA AF145029.1 - 20, 21 2809
AJ133769.1 - 20, 21 2594
AK001263.1 - 13, 14 1551
AK225999.1 - 19, 20 2776
AK312580.1 - 20, 21 2724
BC009221.2 - 8, 9 841
BC068530.1 - 20, 21 2725

View Full Product Details