Gene Transcripts
- Gene Symbol
- POU2F3
- Entrez Gene ID
- 25833
- Gene Name
- POU class 2 homeobox 3
- Gene Aliases
- Epoc-1, OCT-11, OCT11, OTF-11, PLA-1, PLA1, Skn-1a
- Chromosome Location
- Chr. 11: 120236640 - 120319944 on Build GRCh38
- UniGene ID
- Hs.227115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF133895.1 |
- |
3 |
127 |
|
View Full Product Details