Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001307936.1 | NP_001294865.1 | 15 | 2997 | |
NM_018976.4 | NP_061849.2 | 16 | 3113 | ||
GenBank mRNA | AB037803.2 | - | 15 | 2674 | |
AF116620.2 | - | 1 | 663 | ||
AF259799.1 | - | 15 | 2693 | ||
AF298897.1 | - | 16 | 3025 | ||
AJ344099.1 | - | 16 | 3028 | ||
AK001700.1 | - | 15 | 2907 | ||
AK172784.1 | - | 9 | 2120 | ||
BC040342.1 | - | 16 | 3015 | 3874551 |