Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001165960.1 | NP_001159432.1 | 16 | 2883 | |
NM_021628.2 | NP_067641.2 | 16 | 2719 | ||
GenBank mRNA | AF182218.1 | - | 15 | 2550 | |
AJ269499.1 | - | 15 | 2848 | ||
AK296416.1 | - | 16 | 2784 | ||
BC101938.1 | - | 15 | 2571 | 40011855 | |
BC101939.1 | - | 15 | 2571 | 40011856 | |
BC103508.1 | - | 15 | 2571 | 40011857 | |
BC104724.1 | - | 15 | 2571 | 40011849 |