Gene Transcripts
- Gene Symbol
- HARS
- Entrez Gene ID
- 3035
- Gene Name
- histidyl-tRNA synthetase
- Gene Aliases
- CMT2W, HRS, USH3B
- Chromosome Location
- Chr. 5: 140673904 - 140691727 on Build GRCh38
- UniGene ID
- Hs.528050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258041.2 |
NP_001244970.1
|
2, 3 |
611 |
|
NM_001289092.1 |
NP_001276021.1
|
2, 3 |
611 |
|
NM_001289094.1 |
NP_001276023.1
|
2, 3 |
524 |
|
NM_002109.5 |
NP_002100.2
|
2, 3 |
611 |
|
GenBank mRNA |
AK000498.1 |
- |
2, 3 |
291 |
|
AK225776.1 |
- |
2, 3 |
261 |
|
AK293154.1 |
- |
2, 3 |
225 |
|
AK293531.1 |
- |
|
273 |
|
AK307669.1 |
- |
2, 3 |
204 |
|
AY995220.1 |
- |
2, 3 |
188 |
|
BC011807.2 |
- |
2, 3 |
218 |
4331277 |
BC080514.1 |
- |
2, 3 |
216 |
2821065 |
X05345.1 |
- |
2, 3 |
265 |
|
Z11518.1 |
- |
2, 3 |
746 |
|
View Full Product Details