Gene Transcripts

Gene Symbol
AGXT
Entrez Gene ID
189
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6
Chromosome Location
Chr. 2: 240868745 - 240879119 on Build GRCh38
UniGene ID
Hs.144567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000030.2 NP_000021.1 11 1563
GenBank mRNA AB292648.1 - 11 1464
AF191687.1 - 3 1220
BC132819.1 - 11 1560 40146841
X53414.1 - 11 1564
X56092.1 - 11 1463

View Full Product Details