Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014256.3 | NP_055071.2 | 3 | 1717 | |
XM_011527626.2 | XP_011525928.1 | 3 | 1676 | ||
GenBank mRNA | AB015630.1 | - | 3 | 1687 | |
AB049585.1 | - | 3 | 1672 | ||
AK075434.1 | - | 4 | 1267 | ||
AK075466.1 | - | 3 | 1690 | ||
AK225421.1 | - | 3 | 1684 | ||
AY358955.1 | - | 3 | 1672 |