Gene Transcripts
- Gene Symbol
- PDHA1
- Entrez Gene ID
- 5160
- Gene Name
- pyruvate dehydrogenase (lipoamide) alpha 1
- Gene Aliases
- PDHA, PDHAD, PDHCE1A, PHE1A
- Chromosome Location
- Chr. X: 19343893 - 19361707 on Build GRCh38
- UniGene ID
- Hs.530331
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000284.3 |
NP_000275.1
|
6, 7 |
753 |
|
NM_001173454.1 |
NP_001166925.1
|
7, 8 |
867 |
|
NM_001173455.1 |
NP_001166926.1
|
6, 7 |
774 |
|
GenBank mRNA |
AK129887.1 |
- |
6, 7 |
688 |
|
AK222740.1 |
- |
6, 7 |
684 |
|
AK222988.1 |
- |
6, 7 |
718 |
|
AK293250.1 |
- |
7, 8 |
832 |
|
AK296341.1 |
- |
6, 7 |
740 |
|
AK312263.1 |
- |
6, 7 |
719 |
|
BC002406.1 |
- |
6, 7 |
665 |
2961286 |
EF576990.1 |
- |
3, 4 |
222 |
|
EF590117.1 |
- |
|
785 |
|
J03503.1 |
- |
6, 7 |
680 |
|
J03575.1 |
- |
6, 7 |
653 |
|
L13318.1 |
- |
6, 7 |
713 |
|
L48690.1 |
- |
6, 7 |
713 |
|
M24848.1 |
- |
6, 7 |
651 |
|
X52709.1 |
- |
6, 7 |
662 |
|
X52710.1 |
- |
6, 7 |
664 |
|
View Full Product Details