Gene Transcripts

Gene Symbol
MKI67
Entrez Gene ID
4288
Gene Name
marker of proliferation Ki-67
Gene Aliases
KIA, MIB-, MIB-1, PPP1R105
Chromosome Location
Chr. 10: 128096659 - 128126405 on Build GRCh38
UniGene ID
Hs.689823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145966.1 NP_001139438.1 3, 4 371
NM_002417.4 NP_002408.3 3, 4 371
GenBank mRNA X65550.1 - 3, 4 372
X65551.1 - 3, 4 372

View Full Product Details