Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003463.4 | NP_003454.1 | 6 | 1653 | |
XM_011536111.1 | XP_011534413.1 | 6 | 992 | ||
XM_011536112.1 | XP_011534414.1 | 6 | 1751 | ||
XM_017011270.1 | XP_016866759.1 | 6 | 1009 | ||
XM_017011271.1 | XP_016866760.1 | 6 | 989 | ||
GenBank mRNA | AB451314.1 | - | 5 | 500 | |
AB451444.1 | - | 5 | 500 | ||
AK312526.1 | - | 6 | 1137 | ||
BC023975.2 | - | 6 | 1095 | 2960001 | |
BC045571.1 | - | 6 | 1653 | 4826233 | |
CR749458.1 | - | 8 | 1487 | ||
U48296.1 | - | 5 | 892 |