Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
6909
Gene Name
T-box 2
Gene Aliases
Chromosome Location
Chr. 17: 61399896 - 61409466 on Build GRCh38
UniGene ID
Hs.531085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005994.3 NP_005985.3 7 3163
XM_011525159.1 XP_011523461.1 6 2834
GenBank mRNA BC070054.1 - 7 3129

View Full Product Details