Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206804.1 | NP_001193733.1 | 14 | 932 | |
NM_002757.3 | NP_002748.1 | 14 | 1516 | ||
NM_145160.2 | NP_660143.1 | 14 | 1516 | ||
GenBank mRNA | AK024766.1 | - | 3 | 110 | |
AK293459.1 | - | 14 | 932 | ||
AK315435.1 | - | 14 | 1063 | ||
BC008838.2 | - | 14 | 1173 | 4100679 | |
BT006780.1 | - | 14 | 863 | ||
CR542229.1 | - | 14 | 863 | ||
U71087.1 | - | 14 | 924 | ||
U71088.2 | - | 14 | 868 |