Gene Transcripts
- Gene Symbol
- SPATA5
- Entrez Gene ID
- 166378
- Gene Name
- spermatogenesis associated 5
- Gene Aliases
- AFG2, EHLMRS, SPAF
- Chromosome Location
- Chr. 4: 122923070 - 123319450 on Build GRCh38
- UniGene ID
- Hs.709648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317799.1 |
NP_001304728.1
|
6, 7 |
1352 |
|
NM_145207.2 |
NP_660208.2
|
6, 7 |
1355 |
|
XM_005262783.4 |
XP_005262840.1
|
6, 7 |
1352 |
|
XM_011531678.2 |
XP_011529980.1
|
6, 7 |
1352 |
|
XM_011531679.2 |
XP_011529981.1
|
6, 7 |
1310 |
|
XM_017007825.1 |
XP_016863314.1
|
6, 7 |
1351 |
|
XM_017007826.1 |
XP_016863315.1
|
6, 7 |
1351 |
|
XM_017007827.1 |
XP_016863316.1
|
6, 7 |
1351 |
|
XM_017007828.1 |
XP_016863317.1
|
6, 7 |
1317 |
|
XM_017007829.1 |
XP_016863318.1
|
4, 5 |
1975 |
|
XM_017007830.1 |
XP_016863319.1
|
6, 7 |
1351 |
|
GenBank mRNA |
AF361489.1 |
- |
6, 7 |
1351 |
|
AF479656.1 |
- |
6, 7 |
1325 |
|
AK091384.1 |
- |
6, 7 |
1310 |
|
AL832175.1 |
- |
6, 7 |
1348 |
|
BC048217.1 |
- |
6, 7 |
1295 |
6052305 |
View Full Product Details