Gene Transcripts

Gene Symbol
ADCY1
Entrez Gene ID
107
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, DFNB44
Chromosome Location
Chr. 7: 45574140 - 45723116 on Build GRCh38
UniGene ID
Hs.192215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281768.1 NP_001268697.1 7, 8 1018
NM_021116.2 NP_066939.1 6, 7 1332
XM_005249584.3 XP_005249641.1 6, 7 1330
XM_005249585.2 XP_005249642.1 6, 7 1330
GenBank mRNA AB209496.1 - 6, 7 1277
L05500.1 - 4, 5 476

View Full Product Details