Gene Transcripts
- Gene Symbol
- TUBD1
- Entrez Gene ID
- 51174
- Gene Name
- tubulin delta 1
- Gene Aliases
- TUBD
- Chromosome Location
- Chr. 17: 59859467 - 59892948 on Build GRCh38
- UniGene ID
- Hs.463638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193610.1 |
NP_001180539.1
|
6, 7 |
1230 |
|
NM_001193613.1 |
NP_001180542.1
|
3, 4 |
654 |
|
NM_016261.3 |
NP_057345.2
|
6, 7 |
1230 |
|
XM_005257425.1 |
XP_005257482.1
|
5, 6 |
1009 |
|
XM_017024716.1 |
XP_016880205.1
|
6, 7 |
1228 |
|
GenBank mRNA |
AB030837.1 |
- |
5, 6 |
943 |
|
AF201333.1 |
- |
5, 6 |
942 |
|
AK022771.1 |
- |
7, 8 |
1120 |
|
AK024741.1 |
- |
3, 4 |
490 |
|
AK298492.1 |
- |
6, 7 |
1143 |
|
AK301312.1 |
- |
4, 5 |
602 |
|
View Full Product Details