Gene Transcripts

Gene Symbol
IGFBP7
Entrez Gene ID
3490
Gene Name
insulin like growth factor binding protein 7
Gene Aliases
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF
Chromosome Location
Chr. 4: 57031071 - 57110385 on Build GRCh38
UniGene ID
Hs.479808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001553.2 NP_001544.1 5 877
GenBank mRNA BC017201.2 - 5 865
BC066339.1 - 5 877 5296697
L19182.1 - 5 857
S75725.1 - 5 865

View Full Product Details