Gene Transcripts
- Gene Symbol
- GULP1
- Entrez Gene ID
- 51454
- Gene Name
- GULP, engulfment adaptor PTB domain containing 1
- Gene Aliases
- CED-6, CED6, GULP
- Chromosome Location
- Chr. 2: 188291669 - 188595926 on Build GRCh38
- UniGene ID
- Hs.470887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252668.1 |
NP_001239597.1
|
9, 10 |
1287 |
|
NM_001252669.1 |
NP_001239598.1
|
6, 7 |
978 |
|
NM_016315.3 |
NP_057399.1
|
9, 10 |
1287 |
|
NR_045562.1 |
|
9, 10 |
1239 |
|
NR_045563.1 |
|
10, 11 |
1501 |
|
XM_006712589.3 |
XP_006712652.1
|
9, 10 |
1247 |
|
XM_011511335.2 |
XP_011509637.1
|
8, 9 |
1109 |
|
XM_017004306.1 |
XP_016859795.1
|
9, 10 |
1619 |
|
XM_017004307.1 |
XP_016859796.1
|
8, 9 |
1152 |
|
XM_017004308.1 |
XP_016859797.1
|
8, 9 |
1109 |
|
GenBank mRNA |
AB209890.1 |
- |
1, 2 |
92 |
|
AF191771.1 |
- |
9, 10 |
1046 |
|
AF200715.1 |
- |
9, 10 |
1052 |
|
AK023668.1 |
- |
9, 10 |
1365 |
|
AK055718.1 |
- |
9, 10 |
1287 |
|
AK298626.1 |
- |
6, 7 |
607 |
|
AK314498.1 |
- |
9, 10 |
1081 |
|
View Full Product Details