Gene Transcripts

Gene Symbol
LIN7C
Entrez Gene ID
55327
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Gene Aliases
LIN-7-C, LIN-7C, MALS-3, MALS3, VELI3
Chromosome Location
Chr. 11: 27494418 - 27506779 on Build GRCh38
UniGene ID
Hs.91393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018362.3 NP_060832.1 5 519
GenBank mRNA AK002077.1 - 5 513
AL833601.1 - 4 438
BC009774.1 - 5 506
BC053907.1 - 5 496 6502685
DA231006.1 - 5 519

View Full Product Details