Gene Transcripts
- Gene Symbol
- RNASEH1
- Entrez Gene ID
- 246243
- Gene Name
- ribonuclease H1
- Gene Aliases
- H1RNA, PEOB2, RNH1
- Chromosome Location
- Chr. 2: 3531813 - 3558371 on Build GRCh38
- UniGene ID
- Hs.568006
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286834.1 |
NP_001273763.1
|
2, 3 |
363 |
|
NM_001286837.1 |
NP_001273766.1
|
2, 3 |
601 |
|
NM_002936.4 |
NP_002927.2
|
2, 3 |
363 |
|
GenBank mRNA |
AF039652.1 |
- |
2, 3 |
333 |
|
AF048994.1 |
- |
2, 3 |
279 |
|
AF048995.1 |
- |
2, 3 |
313 |
|
AJ224117.1 |
- |
2, 3 |
252 |
|
AK075490.1 |
- |
2, 3 |
312 |
|
AK096913.1 |
- |
2, 3 |
519 |
|
BC002973.1 |
- |
2, 3 |
322 |
3537074 |
BT019670.1 |
- |
2, 3 |
252 |
|
CR541820.1 |
- |
2, 3 |
252 |
|
DA532761.1 |
- |
2, 3 |
363 |
|
View Full Product Details