Gene Transcripts
- Gene Symbol
- ALDOA
- Entrez Gene ID
- 226
- Gene Name
- aldolase, fructose-bisphosphate A
- Gene Aliases
- ALDA, GSD12, HEL-S-87p
- Chromosome Location
- Chr. 16: 30053090 - 30070420 on Build GRCh38
- UniGene ID
- Hs.513490
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000034.3 |
NP_000025.1
|
11, 12 |
1704 |
|
NM_001127617.2 |
NP_001121089.1
|
6, 7 |
926 |
|
NM_001243177.1 |
NP_001230106.1
|
7, 8 |
1047 |
|
NM_184041.2 |
NP_908930.1
|
6, 7 |
893 |
|
NM_184043.2 |
NP_908932.1
|
6, 7 |
865 |
|
XM_011545768.2 |
XP_011544070.1
|
8, 9 |
1567 |
|
GenBank mRNA |
AK098778.1 |
- |
9, 10 |
1745 |
|
AK301993.1 |
- |
7, 8 |
859 |
|
BC000367.2 |
- |
3, 4 |
305 |
|
BC004333.1 |
- |
11, 12 |
1624 |
3630974 |
BC010660.1 |
- |
6, 7 |
811 |
3869603 |
BC012880.1 |
- |
6, 7 |
739 |
3459009 |
BC013614.1 |
- |
6, 7 |
822 |
3880857 |
BC015888.2 |
- |
6, 7 |
735 |
3911623 |
BC016170.1 |
- |
3, 4 |
305 |
|
BC016800.1 |
- |
7, 8 |
763 |
4072487 |
BU855133.1 |
- |
1, 2 |
12 |
|
BX647566.1 |
- |
6, 7 |
715 |
|
CR536528.1 |
- |
5, 6 |
617 |
|
CR541880.1 |
- |
5, 6 |
617 |
|
EF036496.1 |
- |
3, 4 |
265 |
|
EF036497.1 |
- |
2, 3 |
196 |
|
FJ474908.1 |
- |
7, 8 |
784 |
|
M11560.1 |
- |
7, 8 |
784 |
|
M21190.1 |
- |
2, 3 |
203 |
|
X05236.1 |
- |
6, 7 |
763 |
|
View Full Product Details