Gene Transcripts

Gene Symbol
BOLA1
Entrez Gene ID
51027
Gene Name
bolA family member 1
Gene Aliases
CGI-143
Chromosome Location
Chr. 1: 149899582 - 149900795 on Build GRCh38
UniGene ID
Hs.13880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321025.1 NP_001307954.1 2 962
NM_001321026.1 NP_001307955.1 2 1020
NM_016074.4 NP_057158.1 2 795
XM_006711348.3 XP_006711411.1 2 828
GenBank mRNA AF151901.1 - 2 731
BC002771.2 - 2 755
BC063405.1 - 2 755 5190565
BQ277416.1 - 1 161

View Full Product Details