Gene Transcripts

Gene Symbol
ANKRD45
Entrez Gene ID
339416
Gene Name
ankyrin repeat domain 45
Gene Aliases
CT117
Chromosome Location
Chr. 1: 173608336 - 173705430 on Build GRCh38
UniGene ID
Hs.130054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198493.2 NP_940895.1 6 1685
GenBank mRNA AK127170.1 - 6 1685

View Full Product Details