Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010938.1 | NP_001010938.1 | 3 | 556 | |
NM_001308046.1 | NP_001294975.1 | 3 | 614 | ||
NM_005781.4 | NP_005772.3 | 3 | 765 | ||
XM_005269268.3 | XP_005269325.1 | 3 | 556 | ||
XM_005269270.3 | XP_005269327.1 | 3 | 404 | ||
XM_005269275.3 | XP_005269332.1 | 3 | 578 | ||
XM_011512317.2 | XP_011510619.1 | 5 | 1123 | ||
XM_011512318.2 | XP_011510620.2 | 3 | 495 | ||
XM_011512320.1 | XP_011510622.1 | 3 | 278 | ||
XM_017005508.1 | XP_016860997.1 | 3 | 495 | ||
XM_017005509.1 | XP_016860998.1 | 3 | 495 | ||
XM_017005510.1 | XP_016860999.1 | 3 | 495 | ||
GenBank mRNA | AK131539.1 | - | 3 | 556 | |
AK225786.1 | - | 3 | 561 | ||
BC008884.2 | - | 3 | 411 | ||
BC020389.1 | - | 3 | 605 | ||
BC028164.1 | - | 3 | 279 | ||
DQ596952.1 | - | 1 | 14 | ||
L13738.2 | - | 3 | 840 |