Gene Transcripts

Gene Symbol
ANKRD39
Entrez Gene ID
51239
Gene Name
ankyrin repeat domain 39
Gene Aliases
Chromosome Location
Chr. 2: 96847985 - 96858019 on Build GRCh38
UniGene ID
Hs.709507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016466.5 NP_057550.3 4 733
GenBank mRNA AF151034.1 - 4 714
BC031303.2 - 4 725 5284947

View Full Product Details