Gene Transcripts

Gene Symbol
EMC3
Entrez Gene ID
55831
Gene Name
ER membrane protein complex subunit 3
Gene Aliases
POB, TMEM111
Chromosome Location
Chr. 3: 9962680 - 10011248 on Build GRCh38
UniGene ID
Hs.475392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018447.3 NP_060917.1 4, 5 767
XM_005265321.3 XP_005265378.1 4, 5 635
GenBank mRNA AF157321.1 - 3, 4 479
AK172843.1 - 3, 4 475
AK222953.1 - 3, 4 477
AK312008.1 - 3, 4 474
BC022807.1 - 3, 4 462 4897577

View Full Product Details