Gene Transcripts

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
Chromosome Location
Chr. 19: 35758139 - 35770115 on Build GRCh38
UniGene ID
Hs.527982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039887.2 NP_001034976.2 11 1845
XM_011526528.1 XP_011524830.1 11 2393
XM_011526529.2 XP_011524831.1 11 2404
XM_011526530.1 XP_011524832.1 11 2372
XM_011526531.1 XP_011524833.1 11 2360
XM_011526532.1 XP_011524834.1 11 2227
XM_011526533.2 XP_011524835.1 11 2588
XM_011526534.1 XP_011524836.1 12 2393
XM_011526535.2 XP_011524837.1 9 2100
GenBank mRNA AK055219.1 - 10 2614
BC110893.1 - 10 1775 3914279

View Full Product Details