Gene Transcripts

Gene Symbol
C3orf58
Entrez Gene ID
205428
Gene Name
chromosome 3 open reading frame 58
Gene Aliases
DIA1, GoPro49, HASF
Chromosome Location
Chr. 3: 143971798 - 143992368 on Build GRCh38
UniGene ID
Hs.288954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134470.1 NP_001127942.1 3 1732
NM_173552.3 NP_775823.1 3 2686
XM_011512543.2 XP_011510845.1 2 2271
XM_017005869.1 XP_016861358.1 3 1566
GenBank mRNA AK025119.1 - 1 294
AK095063.1 - 1 1051
AK095161.1 - 3 1748
AK095400.1 - 3 1593
AL832839.1 - 3 1747
BC037293.2 - 3 2413 5267770

View Full Product Details