Gene Transcripts

Gene Symbol
FAM206A
Entrez Gene ID
54942
Gene Name
family with sequence similarity 206 member A
Gene Aliases
C9orf6, CG-8, Simiate
Chromosome Location
Chr. 9: 108934393 - 108950745 on Build GRCh38
UniGene ID
Hs.29276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017832.3 NP_060302.1 6 619
GenBank mRNA AF153417.1 - 6 549
AK000464.1 - 6 603
AK310297.1 - 5 445
BC015795.1 - 6 831 3910513

View Full Product Details